missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rabbit Serum

Catalog No. 16120099
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16120099 100 mL
16120107 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16120099 Supplier Gibco™ Supplier No. 16120099

Standard (sterile-filtered)

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Sera Category: Specialty
  • Origin: United States
  • Age: 6 months or less

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Rabbit
Age 6 months or less
Country of Origin United States
Form Liquid
Product Type Rabbit Serum
Quantity 100 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Product Line Gibco
Shipping Condition Frozen
Show More Show Less
What species of rabbit does the Rabbit Serum (Cat. No. 16120099) come from?

The rabbit species is Oryctolagus cuniculus.

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.