missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Gentamicin (50 mg/mL)
Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea
$148.00 - $1,304.00
Specifications
| Type | Gentamicin |
|---|---|
| Physical Form | Liquid |
| Sterile Filtered | Yes |
| Concentration | 50 mg/mL |
| Validated Application | Prevention of Cell Culture Contamination |
Description
- Gentamicin sulfate is a water-soluble antibiotic originally purified from the fungus Micromonospora purpurea
- It acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria
- Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used to prevent the contamination of cell cultures by bacteria
- The recommended working concentration ranges from 0.5 to 50 μg /mL
- Store at 15°C to 30°C
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
Specifications
| Gentamicin | |
| Yes | |
| Prevention of Cell Culture Contamination |
| Liquid | |
| 50 mg/mL | |
| Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.