missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gentamicin (50 mg/mL)

Catalog No. 15750060
Encompass
Change view
Click to view available options
Quantity:
10 mL
10 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15750060 10 mL
15750078 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15750060 Supplier Gibco™ Supplier No. 15750060

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea

  • Gentamicin sulfate is a water-soluble antibiotic originally purified from the fungus Micromonospora purpurea
  • It acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria
  • Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used to prevent the contamination of cell cultures by bacteria
  • The recommended working concentration ranges from 0.5 to 50 μg /mL
  • Store at 15°C to 30°C

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Type Gentamicin
Physical Form Liquid
Sterile Filtered Yes
Concentration 50 mg/mL
Quantity 10 mL
Validated Application Prevention of Cell Culture Contamination
Shipping Condition Room Temperature
Once Gentamicin (50 mg/mL; Cat. No. 15750060) is added to the cell culture medium, can this resulting medium, including gentamicin, be frozen and thawed before using it in cell culture without affecting the performance of the antibiotic?

Yes, gentamicin can be frozen in a prepared medium. However, we don't have any stability data on this process with media not made here in our facility. So, you would have to determine the effectiveness of the product upon thawing empirically.

How can I decontaminate my cultures?

When an irreplaceable culture becomes contaminated, researchers may attempt to eliminate or control the contamination.

1. Determine if the contamination is bacteria, fungus, mycoplasma, or yeast. Read more here to view characteristics of each contaminant.
2. Isolate the contaminated culture from other cell lines.
3. Clean incubators and laminar flow hoods with a laboratory disinfectant, and check HEPA filters.
4. Antibiotics and antimycotics at high concentrations can be toxic to some cell lines. Therefore, perform a dose-response test to determine the level at which an antibiotic or antimycotic becomes toxic. This is particularly important when using an antimycotic such as Gibco Fungizone reagent or an antibiotic such as tylosin.

The following is a suggested procedure for determining toxicity levels and decontaminating cultures:

1. Dissociate, count, and dilute the cells in antibiotic-free media. Dilute the cells to the concentration used for regular cell passage.
2. Dispense the cell suspension into a multiwell culture plate or several small flasks. Add the antibiotic of choice to each well in a range of concentrations. For example, we suggest the following concentrations for Gibco Fungizone reagent: 0.25, 0.50, 1.0, 2.0, 4.0, and 8.0 µg/mL.
3. Observe the cells daily for signs of toxicity such as sloughing, appearance of vacuoles, decrease in confluency, and rounding.
4. When the toxic antibiotic level has been determined, culture the cells for two to three passages using the antibiotic at a concentration one- to two-fold lower than the toxic concentration.
5. Culture the cells for one passage in antibiotic-free media.
6. Repeat step 4.
7. Culture the cells in antibiotic-free medium for four to six passages to determine if the contamination has been eliminated.

What antibiotics do you offer to help control or eliminate cell culture contamination?

Please view the following page to browse the cell culture antibiotics we offer (https://www.thermofisher.com/us/en/home/life-science/cell-culture/mammalian-cell-culture/antibiotics.html).

For Research Use Only. Not for use in diagnostic procedures.