missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HBSS, no calcium, no magnesium, no phenol red DFS Item

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

$65.25 - $562.00

Specifications

Chemical Name or Material Hanks' Balanced Salt Solution (HBSS)
Color Clear
Concentration 1 X
For Use With (Application) Mammalian Cell Culture
pH 6.7 - 7.8
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
14175095 DFS Item
View Documents
Gibco™
14175095
500 mL
Each for $65.25
Only null left
 
14175103 DFS Item
View Documents
Gibco™
14175103
10 x 500 mL
Each for $562.00
Only null left
 
14175079 DFS Item
View Documents
Gibco™
14175079
1000 mL
Each for $110.00
Only null left
 
14175145 DFS Item
View Documents
Gibco™
14175145
6 x 1000 mL
Each for $377.00
Only null left
 
Description

Description

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.

This HBSS is modified as follows:

With: Glucose

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Hanks' Balanced Salt Solution (HBSS)
1 X
6.7 - 7.8
Gibco
Sterile-filtered
cGMP-compliant under the ISO 13485 standard
Clear
Mammalian Cell Culture
Liquid
Hank's Balanced Salt
No Sodium Pyruvate
270 - 295 mOsm/kg
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.