Learn More
Gibco™ HBSS, no calcium, no magnesium, no phenol red

Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.
This HBSS is modified as follows:
With: Glucose
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | Hanks' Balanced Salt Solution (HBSS) |
| Color | Clear |
| Concentration | 1 X |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 6.7 - 7.8 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 6 x 1000 mL |
| Solution Type | Hank's Balanced Salt |
| Sterility | Sterile-filtered |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.