Learn More
Gibco™ DPBS (10X), calcium, magnesium

Description
Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
This DPBS is modified as follows:
With: Calcium, Magnesium
Without: Phenol Red
The complete formulation is available.
Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Concentration | 10 X |
| For Use With (Application) | Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue; Diluting Cells for Counting; and Preparing Reagents |
| pH | 4.4 - 4.7 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 500 mL |
| Solution Type | Dulbecco's Phosphate Buffered Saline |
| Sterility | Sterile-filtered |
| Includes | No Sodium Pyruvate |
| Show More |
Frequently Asked Questions (FAQs)
Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.
Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.