missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bovine Serum, heat inactivated, New Zealand origin
GREENER_CHOICE

Collected from cattle typically 12 months or more in age.

$63.50 - $203.00

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age ≥ 12 months
Country of Origin New Zealand
Form Liquid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
26170035
Encompass
View Documents Promotion Details
Gibco™
26170035
100 mL
Each for $63.50
Only null left
 
26170043
GREENER_CHOICE
Encompass
View Documents Promotion Details
Gibco™
26170043
500 mL
Each for $203.00
Only null left
 
Description

Description

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Origin: New Zealand.

Collected from cattle typically 12 months or more in age.

Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
≥ 12 months
Liquid
Adult Bovine Serum
Sterile-filtered
Gibco
Cattle/Bovine
New Zealand
Bottle
Heat-inactivated
Sterile-filtered
Frozen
Videos
SDS
Documents

Documents

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.