missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bovine Serum, heat inactivated, New Zealand origin

Catalog No. 26170035
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26170035 100 mL
26170043 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26170035 Supplier Gibco™ Supplier No. 26170035
Season of Savings
View all promotions

missing translation for 'validMainframeMsg'

Collected from cattle typically 12 months or more in age.

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Origin: New Zealand.

Collected from cattle typically 12 months or more in age.

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age ≥ 12 months
Country of Origin New Zealand
Form Liquid
Format Bottle
Product Type Adult Bovine Serum
Quantity 100 mL
Serum Treatment Heat-inactivated
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Product Line Gibco
Shipping Condition Frozen
Show More Show Less
When should I use heat-inactivated serum?

Heat-inactivation of serum inactivates the “complements” in the serum. In general, it is not necessary to use heat-inactivated serum to culture most cell types. We recommend using heat-inactivated serum when working with immune type cells or for immunological applications.

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.