Learn More
Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles
GREENER_CHOICE
Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.
$76.86 - $95.84
Specifications
Capacity (Metric) 500 mL
Material LDPE
Mouth Wide
Closure Material Polypropylene
Closure Size 53 mm
View More Specs
Products 6
Catalog Number Mfr. No. Chemical Label Price Quantity & Availability  
Catalog Number Mfr. No. Chemical Label Price Quantity & Availability  
FB11865150
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FB11865150
Acetone
Pack of 3 Each for $77.05
Case of 8 Pack for $616.40
Only null left
FB11865163
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FB11865163
Ethanol
Pack of 3 Each for $95.05
Case of 8 Pack for $760.40
Only null left
FB11865155
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FB11865155
Isopropanol
Pack of 3 Each for $76.86
Case of 8 Pack for $614.88
Only null left
FB11865157
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FB11865157
Methanol
Pack of 3 Each for $82.98
Case of 8 Pack for $663.84
Only null left
FB11865160
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FB11865160
Sodium Hypochlorite
Pack of 3 Each for $80.32
Case of 8 Pack for $642.56
Only null left
FB11865164
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FB11865164
Assorted
Pack of 4 Each for $95.84
Case of 6 Pack for $575.04
Only null left
Description
Description
  • Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Wide mouth 53mm (2 in.) for easy refilling
  • Color-coded polypropylene closures aid in quick and easy identification
  • Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
    Bottle with Label: TOLUENE
    • Red LDPE bottle
    • Does not include vented valve
    Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
    • Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word
    Bottle with Label: METHYL ETHYL KETONE
    • Does not include vented valve
    Compliance: Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to “Workplace Labels”.
Specifications
Specifications
500 mL
Wide
53 mm
Not Autoclavable
LDPE
Polypropylene
Cap