missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles
GREENER_CHOICE

Catalog No. FB11865164
Change view
Click to view available options
Chemical Label:
Acetone
Assorted
Ethanol
Isopropanol
Methanol
Sodium Hypochlorite
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Label
FB11865164 Assorted
FB11865150 Acetone
FB11865163 Ethanol
FB11865155 Isopropanol
FB11865157 Methanol
FB11865160 Sodium Hypochlorite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. FB11865164 Supplier Fisherbrand™ Supplier No. FB11865164
Only null left

Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.

  • Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Wide mouth 53mm (2 in.) for easy refilling
  • Color-coded polypropylene closures aid in quick and easy identification
  • Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol

     

    Bottle with Label: TOLUENE

    • Red LDPE bottle
    • Does not include vented valve

     

    Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
    • Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word

     

    Bottle with Label: METHYL ETHYL KETONE
    • Does not include vented valve

    Compliance: Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to “Workplace Labels”.

Specifications

Capacity (Metric) 500 mL
Color Assorted
Material LDPE
Quantity 4/Bag
Closure Color Assorted
Mouth Wide
Chemical Label Assorted
Closure Material Polypropylene
Closure Size 53 mm
Closure Type Cap
Autoclavable Not Autoclavable
Show More Show Less