missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles
GREENER_CHOICE

Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.

Supplier:  Fisherbrand™ FB11865157

 View more versions of this product

Catalog No. FB11865157



Only null left

Description

Description

  • Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Wide mouth 53mm (2 in.) for easy refilling
  • Color-coded polypropylene closures aid in quick and easy identification
  • Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol

     

    Bottle with Label: TOLUENE

    • Red LDPE bottle
    • Does not include vented valve

     

    Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
    • Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word

     

    Bottle with Label: METHYL ETHYL KETONE
    • Does not include vented valve

    Compliance: Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to “Workplace Labels”.

Specifications

Chemical Identifiers

500 mL
Wide
53 mm
Not Autoclavable
LDPE
Polypropylene
Cap

Specifications

500 mL
LDPE
Green
Methanol
53 mm
Not Autoclavable
Natural
3/Bag
Wide
Polypropylene
Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions