missing translation for 'onlineSavingsMsg'
Learn More

GHRHR Polyclonal Antibody, Invitrogen™

Numéro de catalogue. PIPA120734
Modifier l'affichage
Click to view available options
Quantité:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIPA120734 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA120734 Fournisseur Thermo Fisher Scientific Code fournisseur PA120734

Rabbit Polyclonal Antibody

PA1-20734 detects GHRHR in Human samples.

Growth Hormone Releasing Hormone (GHRH) is a member of a family of peptide hormones that also includes VIP (vasoactive intestinal polypeptide) and PACAP (pituitary adenylate cyclase-activating polypeptide). It is a hypothalamic peptide that binds to G-protein coupled receptors to stimulate secretion and synthesis of growth hormone (GH). GHRH receptor proteins have been identified on the plasma membrane of primary human tumor cells, and are thought to be involved in the growth of a variety of cancer cells.

Spécifications

Antigène GHRHR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 0.1% sodium azide
Gène Ghrhr
Numéro d’acquisition du gène Q02643
Alias du gène GHRFR; GHRH receptor; GHRHR; Ghrh-r; GHRHREC; GHRHRpsv; GRF receptor; GRFR; Growth hormone - releasing receptor; growth hormone releasing hormone receptor; growth hormone-releasing factor receptor; Growth hormone-releasing hormone receptor; IGHD1B; lit; little
Symboles de gène Ghrhr
Espèces hôtes Rabbit
Immunogène Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human GHRHR
Méthode de purification Antigen affinity chromatography
Quantité 25 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 2692
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats