Learn More
Bulbtronics™ Germicidal Lamp
Supplier:  Bulbtronics™ 163
Encompass
 Afficher plus de versions de ce produit
Numéro de catalogue. 14395335
Il en reste null
Description
Description
Spécifications
Identifiants chimiques
Germicidal UV-C
254 nm
Linear Double-ended Lamp
Spécifications
Medium Bi-pin (G13)
20 W
1.25 in.
23.1 in.
T10
Germicidal UV-C
254 nm
3.17 cm
58.85 cm
Linear Double-ended Lamp
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS