missing translation for 'onlineSavingsMsg'
Learn More

Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis)

Catalog No. AA0012318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0012318 50 g
AA0012309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0012318 Supplier Thermo Scientific Chemicals Supplier No. 00012318

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-56-4 | Ge | 72.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-56-4
Molecular Formula Ge
Molecular Weight (g/mol) 72.63
MDL Number MFCD00085310
InChI Key GNPVGFCGXDBREM-UHFFFAOYSA-N
Synonym element, germanio, germanium, metal powder, unii-00072j7xws, powder, germanium, chips trace metals basis, atom, copper metal, germanium, disks, 15mm, thickness 1.0mm, polycrystalline, germanium, powder
PubChem CID 6326954
ChEBI CHEBI:30441
IUPAC Name germanium
SMILES [Ge]

Specifications

Quantity 50 g
Odor Odorless
Particle Size 2cm (0.8 in.) & down
Form Pieces
Percent Purity 99.9999%
Assay (metals basis)
Chemical Name or Material Germanium pieces, Electrical Resistivity: 40 ohm-cm
EINECSNumber 231-164-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only