missing translation for 'onlineSavingsMsg'
Learn More

Germanium(IV) methoxide, 98%

Catalog No. AAA1843714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1843714 25 g
AAA1843706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1843714 Supplier Thermo Scientific Chemicals Supplier No. A1843714

CAS: 992-91-6 | C4H12GeO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 992-91-6
Molecular Formula C4H12GeO4
MDL Number MFCD00014881
Synonym tetramethoxygermanium, tetramethoxygermanium iv, methyl germanate iv

Specifications

Boiling Point 145°C
Melting Point -18°C
Quantity 25 g
Linear Formula (CH3O)4Ge
UN Number UN2929
Flash Point 40°C (105°F)
Formula Weight 196.73
Percent Purity 98%
Odor Like Alcohol
Refractive Index 1.4015
Sensitivity Moisture sensitive
Density 1.3254^2^0
Chemical Name or Material Germanium(IV) methoxide
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H318-H335
Hazard Statement GHS H Statement
H330-H226-H315-H319-H335
Fatal if inhaled.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only