missing translation for 'onlineSavingsMsg'
Learn More

Geranylacetone 96.0+%, TCI America™

Catalog No. G023625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G023625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G023625ML Supplier TCI America Supplier No. G023625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 689-67-8
Molecular Formula C13H22O
Molecular Weight (g/mol) 194.318
MDL Number MFCD00008910
InChI Key HNZUNIKWNYHEJJ-FMIVXFBMSA-N
Synonym 6,10-Dimethyl-5,9-undecadien-2-one, Geranyl-2-propanone
PubChem CID 1549778
ChEBI CHEBI:67206
IUPAC Name (5E)-6,10-dimethylundeca-5,9-dien-2-one
SMILES CC(=CCCC(=CCCC(=O)C)C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 194.32
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Geranylacetone [mixture of (E)- and (Z)- isomers, (3:2)]
EINECSNumber (2)-0574
RTECSNumber YQ1190000
TSCA Yes