missing translation for 'onlineSavingsMsg'
Learn More

Geranylacetone, (E)+(Z), 97%, (Z)-isomer (nerylacetone) ca 45%

Catalog No. AAA1918414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1918414 25 g
AAA1918422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1918414 Supplier Thermo Scientific Chemicals Supplier No. A1918414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 689-67-8 | C13H22O | 194.318 g/mol

It is used as a flavoring agent and adjuvant. It is also employed as a substance, extract, or preparation for diffusing or imparting an agreeable or attractive smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a flavoring agent and adjuvant. It is also employed as a substance, extract, or preparation for diffusing or imparting an agreeable or attractive smell.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refregirated.

Chemical Identifiers

CAS 689-67-8
Molecular Formula C13H22O
Molecular Weight (g/mol) 194.318
MDL Number MFCD00008910
InChI Key HNZUNIKWNYHEJJ-FMIVXFBMSA-N
Synonym geranylacetone, geranyl acetone, e-6,10-dimethylundeca-5,9-dien-2-one, trans-geranylacetone, 5e-6,10-dimethylundeca-5,9-dien-2-one, geranylacetone, trans, dihydropseudoionone, 6,10-dimethylundeca-5,9-dien-2-one, e-6,10-dimethyl-5,9-undecadien-2-one, unii-9b7ry79u9z
PubChem CID 1549778
ChEBI CHEBI:67206
IUPAC Name (5E)-6,10-dimethylundeca-5,9-dien-2-one
SMILES CC(=CCCC(=CCCC(=O)C)C)C

Specifications

Density 0.87
Boiling Point 250°C
Flash Point 110°C (230°F)
Refractive Index 1.467
Quantity 25 g
Beilstein 1722277
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 194.32
Percent Purity 97%
Assay (Z)-isomer (nerylacetone) ≈45%
Chemical Name or Material Geranylacetone, (E)+(Z)
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 211-711-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only