missing translation for 'onlineSavingsMsg'
Learn More

Gentian Violet, 2% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 323316

Quantity 500 mL
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. 323316

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

548-62-9 , 7732-18-5
407.986
11057
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
C25H30ClN3
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
CHEBI:41688
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

Gentian Violet
96.0, 1.92
C25H30ClN3
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
407.986
CHEBI:41688
Aqueous Solution
500 mL
548-62-9 , 7732-18-5
100.0, 2.0
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
Natural Poly Bottle
Violet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents