missing translation for 'onlineSavingsMsg'
Learn More

Gentian Violet, 1% (w/v), Ricca Chemical

Chemical Identifiers

CAS 548-62-9 , 7732-18-5
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) Mixture
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
32304
View Documents Promotion Details
N/A 120 mL Natural Poly Bottle N/A
Only null left
 
323016
View Documents Promotion Details
N/A 500 mL Natural Poly Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

548-62-9 , 7732-18-5
Mixture
11057
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
C25H30ClN3
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
CHEBI:41688
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

548-62-9 , 7732-18-5
100.0, 0.71
ZXJXZNDDNMQXFV-UHFFFAOYSA-M
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
11057
1% W/V
Dark Violet
Gentian Violet
97.0, 0.68
C25H30ClN3
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]
Mixture
CHEBI:41688
Indicator
4
Videos
SDS
Documents

Documents