missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentian Violet, 1% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 323016
| Quantity | 500 mL |
|---|---|
| Packaging | Natural Poly Bottle |
Chemical Identifiers
| 548-62-9 , 7732-18-5 | |
| Mixture | |
| 11057 | |
| [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride |
| C25H30ClN3 | |
| ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
| CHEBI:41688 | |
| CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] |
Specifications
| Gentian Violet | |
| 97.0, 0.68 | |
| C25H30ClN3 | |
| CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
| Mixture | |
| CHEBI:41688 | |
| Indicator | |
| Dark Violet | |
| 500 mL |