missing translation for 'onlineSavingsMsg'
Learn More

Gentian Violet, 1% Aqueous, Certified, LabChem™

Catalog No. LC148107
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC148107 125 mL Poly Bottle
LC148101 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC148107 Supplier LabChem Supplier No. LC148107

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 548-62-9 , 7732-18-5
Molecular Formula C25H30ClN3
Molecular Weight (g/mol) 407.986
InChI Key ZXJXZNDDNMQXFV-UHFFFAOYSA-M
PubChem CID 11057
ChEBI CHEBI:41688
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-]

Specifications

CAS Max % 99, 1
Identification Passes Test
Packaging Poly Bottle
Linear Formula C25H30CIN3
Decomposition Information Carbon monoxide; Carbon dioxide; Hydrogen chloride; Nitrogen oxides
Color Purple
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Formula Weight 407.99
Grade Certified
Chemical Name or Material Gentian Violet, 1% Aqueous
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Suspected of causing cancer.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
f exposed or concerned: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature