missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentamicin Sulfate USP, Medisca™

$92.00 - $4,946.81
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
18048032
![]() |
N/A | 5 g |
Each for $92.00
|
|
|||||
18048069
![]() |
N/A | 25 g |
Each for $288.27
|
|
|||||
18048091
![]() |
N/A | 1 kg |
Each for $4,946.81
|
|
|||||
18048127
![]() |
N/A | 100 g |
Each for $907.79
|
|
|||||
18048128
![]() |
N/A | 500 g |
Each for $3,082.18
|
|
|||||
Description
Gentamicin Sulfate USP is a white to buff, hygroscopic powder.
Chemical Identifiers
| 1405-41-0 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 1405-41-0 | |
| ≥ 590 μg of Gentamicin/mg |
| Powder | |
| White to Light Yellow | |
| USP |