Learn More
Gentamicin Sulfate USP, Medisca™
Supplier: Medisca Pharmaceutique Inc 063209
| Quantity | 1 kg |
|---|
Description
Gentamicin Sulfate USP is a white to buff, hygroscopic powder.
Chemical Identifiers
| 1405-41-0 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 1405-41-0 | |
| 1 kg | |
| USP |
| Powder | |
| White to Light Yellow | |
| ≥ 590 μg of Gentamicin/mg |