missing translation for 'onlineSavingsMsg'
Learn More

Gentamicin Sulfate USP, Medisca™
GREENER_CHOICE

Catalog No. 18048128
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048128 500 g
18048032 5 g
18048069 25 g
18048091 1 kg
18048127 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18048128 Supplier Medisca Pharmaceutique Inc Supplier No. 063208

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gentamicin Sulfate USP is a white to buff, hygroscopic powder.

Chemical Identifiers

CAS 1405-41-0

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Powder
Color White to Light Yellow
Quantity 500 g
Percent Purity ≥ 590 μg of Gentamicin/mg
Grade USP