missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amsbio LLC (formerly Genlantis) GeneSilencer™ siRNA Transfection Reagents
Novel cationic lipid formulation specifically designed for efficient delivery of siRNAs (small interfering RNAs) into a wide variety of cell types
$634.75 - $5108.48
Specifications
| For Use With (Application) | Efficient delivery of >80% gene suppression siRNAs (small interfering RNAs) into a wide variety of cell types |
|---|---|
| Includes | Hydrated GeneSilencer™ Lipid, siRNA diluents |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T500020
|
Amsbio LLC (formerly Genlantis)
T500020 |
0.2 mL |
Each for $634.75
|
|
|||||
T500750
|
Amsbio LLC (formerly Genlantis)
T500750 |
0.75 mL |
Each for $1,818.46
|
|
|||||
T505750
|
Amsbio LLC (formerly Genlantis)
T505750 |
5 x 0.75 mL |
Each for $5,108.48
|
|
|||||
Description
- Produces >80% gene suppression in wide range of cell types
- Compatible with or without serum-containing media
- Low cytotoxicity
Specifications
| Efficient delivery of >80% gene suppression siRNAs (small interfering RNAs) into a wide variety of cell types | |
| Hydrated GeneSilencer™ Lipid, siRNA diluents |