missing translation for 'onlineSavingsMsg'
Learn More

Amsbio LLC (formerly Genlantis) GeneSilencer™ siRNA Transfection Reagents DFS Item

Novel cationic lipid formulation specifically designed for efficient delivery of siRNAs (small interfering RNAs) into a wide variety of cell types

Supplier:  Amsbio LLC (formerly Genlantis) T505750

 View more versions of this product

Catalog No. T505750


Only null left
Explore available promotions

Description

Description

  • Produces >80% gene suppression in wide range of cell types
  • Compatible with or without serum-containing media
  • Low cytotoxicity

Specifications

Specifications

Efficient delivery of >80% gene suppression siRNAs (small interfering RNAs) into a wide variety of cell types
Hydrated GeneSilencer™ Lipid, siRNA diluents
5 x 0.75 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions