Learn More
Amsbio LLC (formerly Genlantis) GeneSilencer™ siRNA Transfection Reagents
Novel cationic lipid formulation specifically designed for efficient delivery of siRNAs (small interfering RNAs) into a wide variety of cell types
Supplier: Amsbio LLC (formerly Genlantis) T500750
Description
- Produces >80% gene suppression in wide range of cell types
- Compatible with or without serum-containing media
- Low cytotoxicity
Specifications
| Efficient delivery of >80% gene suppression siRNAs (small interfering RNAs) into a wide variety of cell types | |
| Hydrated GeneSilencer™ Lipid, siRNA diluents | |
| 0.75 mL |