missing translation for 'onlineSavingsMsg'
Learn More

Amsbio LLC (formerly Genlantis) BioPORTER™ Protein Delivery Reagents and Kits DFS Item

Translocate functional proteins directly into a wide range of cell types

$1629.75 - $1852.77

Specifications

Quantity 24 Tests
Content And Storage -20°C
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
BP502424 DFS Item
View Documents
Amsbio LLC (formerly Genlantis)
BP502424
QuikEase Kit
Each for $1,852.77
Only null left
 
BP502401 DFS Item
View Documents
Amsbio LLC (formerly Genlantis)
BP502401
Protein Delivery Reagent
Each for $1,629.75
Only null left
 
Description

Description

  • Efficient delivery of proteins, peptides and other macromolecules into a wide variety of cell types
  • Fast and easy protocol
  • Unique lipid formulation allows direct translocation of proteins into living cells

Specifications

Specifications

24 Tests
-20°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'