Learn More
Abcam Inc Genistein, Protein tyrosine kinase inhibitor, 10MG
Numéro de catalogue. 502707869
Encompass
MW 270.24 Da, Purity >99%. Phytoestrogen found in soy beans. Anticancer agent. Inihibits tyrosine protein kinase and DNA topoisomerase as well as possessing antioxidant and cell cycle inhibitor activity. Endocytosis inhibitor.
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 502707869 Fournisseur Abcam Inc Code fournisseur AB12011210MG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
MW 270.24 Da, Purity >99%. Phytoestrogen found in soy beans. Anticancer agent. Inihibits tyrosine protein kinase and DNA topoisomerase as well as possessing antioxidant and cell cycle inhibitor activity. Endocytosis inhibitor.
The product is subject to the following: Abcam Restricted Use Statement