missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genipin 97.0+%, TCI America™
$108.21 - $330.36
Chemical Identifiers
| CAS | 6902-77-8 |
|---|---|
| Molecular Formula | C11H14O5 |
| Molecular Weight (g/mol) | 226.228 |
| MDL Number | MFCD00888600 |
| InChI Key | AZKVWQKMDGGDSV-BCMRRPTOSA-N |
| Synonym | Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate |
| PubChem CID | 442424 |
| ChEBI | CHEBI:5298 |
| IUPAC Name | methyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate |
| SMILES | COC(=O)C1=COC(C2C1CC=C2CO)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
G045825MG
|
N/A | 25 mg |
Each for $108.21
|
|
|||||
|
G0458100MG
|
N/A | 100 mg |
Each for $330.36
|
|
|||||
Chemical Identifiers
| 6902-77-8 | |
| 226.228 | |
| AZKVWQKMDGGDSV-BCMRRPTOSA-N | |
| 442424 | |
| methyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate |
| C11H14O5 | |
| MFCD00888600 | |
| Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate | |
| CHEBI:5298 | |
| COC(=O)C1=COC(C2C1CC=C2CO)O |
Specifications
| 6902-77-8 | |
| White | |
| MFCD00888600 | |
| Methyl (1S,2R,6S)-2-Hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate | |
| COC(=O)C1=COC(C2C1CC=C2CO)O | |
| 226.228 | |
| CHEBI:5298 | |
| ≥97.0% (GC) | |
| Genipin |
| 121°C | |
| C11H14O5 | |
| 2811 | |
| AZKVWQKMDGGDSV-BCMRRPTOSA-N | |
| methyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1,4a,5,7a-tetrahydrocyclopenta[c]pyran-4-carboxylate | |
| 442424 | |
| 226.23 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : GY5828000
TSCA : No