missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Genipin

Catalog No. AC466640010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466640010 1 g
AC466642500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466640010 Supplier Thermo Scientific Chemicals Supplier No. 466640010

CAS: 6902-77-8 | C11H14O5, C11H14O5 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6902-77-8
Molecular Formula C11H14O5

Specifications

Quantity 1 g
Melting Point 121.0°C
Formula Weight 226.23
InChI Key AZKVWQKMDGGDSV-BCMRRPTOSA-N
SMILES COC(=O)C1=CO[C@@H](O)[C@H]2[C@@H]1CC=C2CO
IUPAC Name methyl (1R,4aS,7aS)-1-hydroxy-7-(hydroxymethyl)-1H,4aH,5H,7aH-cyclopenta[c]pyran-4-carboxylate
Molecular Weight (g/mol) 226.23
Chemical Name or Material Genipin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Causes serious eye irritation.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Store locked up.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only