missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Genetic Analyzer Sample Tubes, 0.5 mL

Catalog No. 401957
Encompass
Change view
Click to view available options
Quantity:
500 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401957 500 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401957 Supplier Applied Biosystems™ Supplier No. 401957

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

500 Tubes

These are sample tubes for use in the 48-sample tray of the 310 Genetic Analyzer.

These are sample tubes for use in the 48-sample tray of the 310 Genetic Analyzer.

Use with: 1.0 mL Glass Syringe, 310 Genetic Analyzer Capillary (47 cm, 36 cm wtr), 310 Running Buffer (10X), BigDye™ Terminator v1.1 Cycle Sequencing Kit, GeneScan™ 500 ROX™ Size Standard, Hi-Di™ Formamide, MicroAmp™ Reaction Tube with Cap (0.2 ml), POP-4™ Polymer for the 310 Genetic Analyzer, POP-6™ Polymer for the 310 Genetic Analyzer, Septa for 0.5 ml Sample Tubes

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.

Specifications

Content And Storage Each package contains 500 tubes.
For Use With (Equipment) 310 Genetic Analyzer
For Use With (Application) Sequencing
Product Type Genetic Analyzer Sample Tube
Quantity 500 Tubes
Shipping Condition Room Temperature

For research use only