missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Buffer

Catalog No. N8080006
Encompass
Change view
Click to view available options
Quantity:
1.5 mL
6 x 1.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N8080006 1.5 mL
N8080129 6 x 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N8080006 Supplier Applied Biosystems™ Supplier No. N8080006

Applied Biosystems GeneAmp 10X PCR Buffer I is optimized for use with AmpliTaq DNA Polymerase, resulting in robust PCR amplification.

Applied Biosystems™ GeneAmp™ 10X PCR Buffer I is optimized for use with AmpliTaq™ DNA Polymerase, resulting in robust PCR amplification.

  • Optimized for use with AmpliTaq™ DNA Polymerase, offering maximum PCR performance
  • Available in several convenient sizes

GeneAmp™ 10X PCR Buffer I contains 15 mM MgCl2. When diluted 1:10, it provides the optimal pH and ionic strength for PCR amplification with AmpliTaq™ DNA Polymerase.

Note: See user's manual or package insert for limited label license and trademark information.

Order Info

Shipping Condition: Dry ice

Specifications

Chemical Name or Material PCR Buffer
Concentration 10X
For Use With (Application) Standard PCR
Quantity 1.5 mL
Includes • GeneAmp 10X PCR Buffer I (with 15 mM MgCl2), 1.5 mL

Store at -15°C to -25°C.
What is BSA and its function in PCR? What concentration of BSA do you recommend?

BSA, bovine serum albumin, is usually used in PCR as a stabilizer. A concentration of 160 µg/mL is typical. Gelatin may also be used as a stabilizer at 0.01% (w/v).

What are the differences between GeneAmp 10X PCR Buffer I, PCR Buffer II and PCR Gold Buffer, and what are the formulations of each buffer?

The ionic strength and the pH of GeneAmp 10X PCR Gold Buffer have been carefully optimized for use with the AmpliTaq Gold enzyme. GeneAmp 10X PCR Buffer I contains Mg; Geneamp 10X PCR Buffer II and Geneamp 10X PCR Gold Buffer do not have Mg in them, but come with a separate vial of 25 mM MgCl2 so that you can titrate the Mg in your PCR reaction.

Buffer formulations:
Buffer I (500 mM KCl, 100 mM Tris-HCl, pH 8.3, 15 mM MgCl2, 0.01% (w/v) gelatin)
Buffer II (500 mM KCl, 100 mM Tris-HCl, pH 8.3), with separate tube of 25 mM MgCl2
Gold Buffer (500 mM KCl, 150 mM Tris-HCl pH 8.0), with separate tube of 25 mM MgCl2

For Research Use Only. Not for use in diagnostic procedures.