missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Gen5 Spot Counting Module Add-On
GREENER_CHOICE

Catalog No. BTGEN5SPOT
Change view
Click to view available options
For Use With (Equipment):
Gen5 Image Prime
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
BTGEN5SPOT Gen5 Image Prime
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTGEN5SPOT Supplier Agilent BioTek Supplier No. GEN5 SPOT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gen5 Spot Counting Module Add-On for Gen5 Image Prime only. Requires Gen5 Image Prime 3.03 or higher.

  • Powerful functionality microplate reading, enabling applications from micro-volume nucleic acid quantification to long-term cell-based kinetic assays.
  • Fluorophore spectra utility offers >100 fluorophore spectra, making filter cube matching quick and easy
  • Enhanced conditional logic adds powerful functionality for data transformation, cutoffs and validation with automated outlier rejection and expanded formula operators
  • Improved file size, sorting and search tools for working with Gen5's database and sample file database

Specifications

Product Type Software
For Use With (Equipment) Gen5 Image Prime