missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorhydrate de gemcitabine 98,0+%, TCI America™
Specifications
Specifications
| Melting Point | 292°C |
| Color | Blanc |
| Quantity | 100 mg |
| Formula Weight | 299.66 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | Gemcitabine Hydrochloride |
Safety and Handling
| RTECSNumber | HA3840000 |
| TSCA | No |