Naturally derived gelling polymer that can be used in place of agar. Gelrite forms clear, rigid gels at approximately half the use level of agar in presence of soluble salts like Mg2+ and Ca2+.
Numéro de catalogue.
50213333
Fournisseur
Research Products CorporationCode fournisseur
RPI G35020250.0
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Naturally derived gelling polymer that can be used in place of agar. Gelrite forms clear, rigid gels at approximately half the use level of agar in presence of soluble salts like Mg2+ and Ca2+.