Learn More
Research Products Corporation Gelrite, 1 Kilogram
Numéro de catalogue. 50488682
Encompass
Naturally derived gelling polymer that can be used in place of agar. Gelrite forms clear, rigid gels at approximately half the use level of agar in presence of soluble salts like Mg2+ and Ca2+.
Numéro de catalogue. 50488682 Fournisseur Research Products Corporation Code fournisseur RPI G350201000.0
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Naturally derived gelling polymer that can be used in place of agar. Gelrite forms clear, rigid gels at approximately half the use level of agar in presence of soluble salts like Mg2+ and Ca2+.