missing translation for 'onlineSavingsMsg'
Learn More

Geldanamycin 98.0+%, TCI America™

Catalog No. G033410MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G033410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G033410MG Supplier TCI America Supplier No. G033410MG

Chemical Identifiers

CAS 30562-34-6
Molecular Formula C29H40N2O9
Molecular Weight (g/mol) 560.644
MDL Number MFCD00274570
InChI Key QTQAWLPCGQOSGP-KDFVLELPSA-N
Synonym geldanamycin nsc122750
PubChem CID 133688416
IUPAC Name [(3S,5R,6S,7R,8Z,11R,14E)-6-hydroxy-5,11,21-trimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate
SMILES CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C(=C(C1)C2=O)OC)C)OC)OC(=O)N)C)C)O)OC

Specifications

Melting Point 255°C
Quantity 10 mg
Formula Weight 560.64
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Geldanamycin
RTECSNumber LX8920000
TSCA No
Recommended Storage Freezer