Electron Microscopy Sciences Gelatin Capsule Size 07
Numéro de catalogue. 5026881
Our large size gelatin capsules are ideal for storing either delicate or small items. It is also suitable for use as an embedding mold for paraffin or paraplast. The capsules come complete with a locking ring.
Numéro de catalogue.
5026881
Fournisseur
Electron Microscopy SciencesCode fournisseur
EMS70118
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Our large size gelatin capsules are ideal for storing either delicate or small items. It is also suitable for use as an embedding mold for paraffin or paraplast. The capsules come complete with a locking ring.