Natural, hard gelatin capsules. To be used as an embedding mold for water miscible resins, or resins which need to be cured by transmitted light. Gelatin capsules dissolve or disintegrate within 5 minutes when immersed in a 0.5% HCl solution at 36-38 degree C, within 2 minutes in water at 37 degree C or in artificial gastric juice.
OAL: 26.14 mm
Diameter: 9.55 mm
Volume: 1.37 mL