Learn More
New England Biolab Ltd Gel Loading Dye Orange (6X) 4 ml
Supplier:  New England Biolab Ltd B7022S
Encompass
Gel Loading Dye, Orange (6X) is a pre-mixed loading buffer with a tracking dye for agarose and non-denaturing polyacrylamide gel electrophoresis. This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to their DNA substrates following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. Orange G will not appear in gel photographs, and runs ahead of all but the smallest restriction fragments. Orange G migrates at approximately 50 bp on a standard 1% TBE agarose gel. This product is packaged as 4x1 ml vials.
Catalog No. 50994906
Only null left