missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GDF7 Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PA5150100
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PA5150100 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PA5150100 Fournisseur Invitrogen™ Code fournisseur PA5150100
Il en reste null

Rabbit Polyclonal Antibody

GDF7 Polyclonal Antibody for Western Blot, ICC/IF, IHC

This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord.

Spécifications

Antigen GDF7
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GDF7
Gene Accession No. P43029
Gene Alias BMP12; Gdf7; Gdf-7; growth differentiation factor 7; growth/differentiation factor 7
Gene Symbols GDF7
Host Species Rabbit
Immunogen Recombinant Growth Differentiation Factor 7 (GDF7) (Ser295-Val447).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238057
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Afficher plus de résultats Afficher moins de résultats