missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, OV-210 Phase

Catalog No. 111002523
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002523 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002523 Supplier MilliporeSigma™ Supelco™ Supplier No. 21240U

Consists of 50% trifluoropropyl groups which generate important electron acceptor effects, and produce an outstanding selectivity

  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase

Specifications

Max. Temperature 275°C
Stationary Phase OV-210
Packaging Bottle
Quantity 25 g
For Use With (Application) Suitable for G6