Learn More
MilliporeSigma™ Supelco™ Gas Sampling Bulbs
For trapping and transporting gas samples for analysis
$350.23 - $465.89
Specifications
For Use With (Application) Trapping and Transporting Gas Samples for Analysis
Type Gas Sampling Bulb
View More Specs
Products 7
Catalog Number Mfr. No. Material Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Material Volume (Metric) Price Quantity & Availability  
111004355
View Documents Promotion Details
MilliporeSigma™ Supelco™
22146U
Glass Stopcock 125 mL
Each for $350.23
Only null left
111004256
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA22161
PTFE Stopcock 125 mL
Each for $394.78
Only null left
111004290
View Documents Promotion Details
MilliporeSigma™ Supelco™
22147U
Glass Stopcock 250 mL
Each for $371.41
Only null left
111004257
View Documents Promotion Details
MilliporeSigma™ Supelco™
22162U
PTFE Stopcock 250 mL
Each for $408.78
Only null left
111004239
View Documents Promotion Details
MilliporeSigma™ Supelco™
22163U
PTFE Stopcock 500 mL
Each for $437.57
Only null left
111004238
View Documents Promotion Details
MilliporeSigma™ Supelco™
22144U
Glass Stopcock 1000 mL
Each for $422.20
Only null left
111016085
View Documents Promotion Details
MilliporeSigma™ Supelco™
22145U
PTFE Stopcock 1000 mL
Each for $465.89
Only null left
Description
Description
  • Use the plug-type septum to easily remove a gas sample for analysis
  • Oven annealed to resist damage during transportation and on-site use
  • Each bulb includes one half-hole, cylindrical Thermogreen™ septum
  • Available with PTFE or high vacuum glass stopcocks
Specifications
Specifications
Trapping and Transporting Gas Samples for Analysis
Gas Sampling Bulb