Learn More
MilliporeSigma™ Supelco™ Gas Sampling Bulbs
For trapping and transporting gas samples for analysis
Supplier:  MilliporeSigma™ Supelco™ 22144U
 View more versions of this product
Catalog No. 111004238
Only null left
Description
Description
  • Use the plug-type septum to easily remove a gas sample for analysis
  • Oven annealed to resist damage during transportation and on-site use
  • Each bulb includes one half-hole, cylindrical Thermogreen™ septum
  • Available with PTFE or high vacuum glass stopcocks
Specifications
Chemical Identifiers
Trapping and Transporting Gas Samples for Analysis
Gas Sampling Bulb
Specifications
Glass Stopcock
Trapping and Transporting Gas Samples for Analysis
Gas Sampling Bulb
1000 mL
Product Suggestions
Product Suggestions