missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GammaBind Plus Sepharose

Catalog No. 45002366
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002366 25 mL
45002365 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002366 Supplier Cytiva Supplier No. 17088602

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Binds to all IgG subclasses. Cytiva GammaBind Plus Sepharose is used for lab-scale affinity purification of IgG from a large variety of species and has better binding to rat and mouse IgG subclasses compared to standard GammaBind Sepharose.

GammaBind Plus Sepharose may be used to analyze and purify classes, subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.
  • Recombinant Protein G type 3 coupled to Sepharose CL-6B for enhanced binding to rat and mouse IgG subclasses at laboratory scale
  • Binds to all IgG subclasses from human, mouse, and rat; binds total IgG from guinea pig, rabbit, goat, cow, sheep, and horse
  • Yields up to 18mg/ml mouse IgG
  • Available in 5ml and 25ml sizes
GammaBind Plus Sepharose is GammaBind G, Type 3, covalently immobilized to Sepharose CL-6B by malimide linkage. GammaBind G, Type 3, a recombinant form of streptococcal protein G, binds to the Fc region of IgG from a variety of mammalian species. Since only the Fc region is involved in binding, the Fab region is still available for binding antigen.

Specifications

Flow Rate >130 cm/h
Ligand Recombinant GammaBind G type 3 lacking albumin-binding region
Particle Size 90 μm, d50v is the median particle size of the cumulative volume distribution.
Content And Storage 2°C to 8°C, 20% Ethanol
Quantity 25 mL