missing translation for 'onlineSavingsMsg'
Learn More

gamma-Octanolactone, 98%

Catalog No. AAA1814214
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1814214 25 g
AAA1814222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1814214 Supplier Thermo Scientific Chemicals Supplier No. A1814214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 104-50-7 | C8H14O2 | 142.198 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-50-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00005402
InChI Key IPBFYZQJXZJBFQ-UHFFFAOYSA-N
Synonym gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone
PubChem CID 7704
IUPAC Name 5-butyloxolan-2-one
SMILES CCCCC1CCC(=O)O1

Specifications

Density 0.981
Boiling Point 233°C to 235°C
Flash Point 126°C (259°F)
Refractive Index 1.444
Quantity 25 g
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material gamma-Octanolactone
Hazard Category H315-H320-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 203-208-1
RTECSNumber LU3562000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only