missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Dodecanolactone 97.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Quantity | 25 mL |
| Formula Weight | 198.31 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | gamma-Dodecanolactone |
Sécurité et manipulation
| EINECSNumber | (9)-0137 |
| RTECSNumber | LU3600000 |
| TSCA | Yes |