missing translation for 'onlineSavingsMsg'
Learn More

gamma-Benzyl L-glutamate, 99%

Catalog No. AC225670250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC225670250 25 g Glass bottle
AC225670050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC225670250 Supplier Thermo Scientific Chemicals Supplier No. 225670250

CAS: 1676-73-9 | C12H15NO4 | 237.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1676-73-9
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.25
MDL Number MFCD00002633
InChI Key BGGHCRNCRWQABU-JTQLQIEISA-N
Synonym h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
PubChem CID 122337
IUPAC Name (2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N

Specifications

Melting Point 162°C to 167°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2O2CCH2CH2CH(NH2)CO2H
Quantity 25 g
Specific Rotation Condition +27.20° (20°C c=2,1N HCL)
Specific Rotation + 27.2
Formula Weight 237.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material γ-Benzyl L-glutamate, 99+%
Show More Show Less
EINECSNumber 216-826-1
TSCA TSCA

RUO – Research Use Only