Learn More
Gallium, plasma standard solution, Specpure™ Ga 1000μg/mL
CAS: 7440-55-3 | Ga | 69.72 g/mol
$128.67 - $128.67
Chemical Identifiers
| CAS | 7440-55-3 |
|---|---|
| Molecular Weight (g/mol) | 69.72 |
| MDL Number | MFCD00134045 |
| InChI Key | GYHNNYVSQQEPJS-UHFFFAOYSA-N |
| PubChem CID | 5360835 |
| ChEBI | CHEBI:49631 |
| IUPAC Name | gallium |
| SMILES | [Ga] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-55-3 | |
| MFCD00134045 | |
| 5360835 | |
| gallium |
| 69.72 | |
| GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
| CHEBI:49631 | |
| [Ga] |
Specifications
| 7440-55-3 , 7647-01-0 , 7732-18-5 | |
| Liquid | |
| MFCD00134045 | |
| [Ga] | |
| 69.72 | |
| 5360835 | |
| Nitric acid: 5.0%; Gallium: 0.1%; Hydrochloric acid: <0.1%; Water: 94.8% |
| 23 hPa (17mm Hg) at 20°C | |
| Ga | |
| GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
| gallium | |
| 100 mL | |
| CHEBI:49631 | |
| Gallium |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8, Packing Group: III
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only